missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD73 Rat anti-Mouse, PerCP-eFluor 710, Clone: eBioTY/11.8 (TY/11.8), eBioscience™

Rat Monoclonal Antibody

$191.40 - $471.25

Specifications

Antigen CD73
Clone eBioTY/11.8 (TY/11.8)
Host Species Rat
Gene Alias Ecto-5'-nucleotidase, NT5E
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50245874 Life Technologies
LS46073182
100μg Each for $471.25
50245873 Life Technologies
LS46073180
25μg Each for $191.40
Specifications

Description

eBioTY/11.8 recognizes the with CD73 a 69kDa GPI-anchored cell-surface protein with ecto-5'-nucleotidase activity. Expression on myeloid cells (CD11b) is restricted to the bone marrow. In human CD73 can be induced to secrete a soluble form with IL-2 suggesting a role in mediating activation signals. Differences between human and mouse CD73 have been reported. BALB/c mice have more CD4+CD73+ than CD8+CD73+ while the reciprocal is documented in humans.

CD73 is expressed on a subset of lymphocytes and increases during lymphocyte maturation. Recently it has been found that memory CD4 T cells express and are similar to the uncommitted primed precursor helper cells (Thpp) that can differentiate into Th1 or Th2 cells. Furthermore CD73 has been found on regulatory T cells.
Description & Specifications

Specifications

CD73
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
eBioTY/11.8 (TY/11.8)
Ecto-5'-nucleotidase, NT5E
Flow Cytometry
PerCP-eFluor 710
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL