missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD73 Mouse anti-Human, PerCP-eFluor 710, Clone: AD2, eBioscience™

Mouse Monoclonal Antibody

$174.24 - $443.52

Specifications

Antigen CD73
Clone AD2
Host Species Murine
Gene Alias Ecto-5'-nucleotidase, NT5E
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5015981 Life Technologies
LS46073941
25 Tests Each for $174.24
5015982 Life Technologies
LS46073942
100 Tests Each for $443.52
Specifications

Description

This AD2 monoclonal antibody reacts with human CD73, a 5’-ectonucleotidase that converts 5’-adenosine monophosphate to adenosine. CD73 is expressed on the surface of endothelial cells, as well as B and T cells, including some CD4+Foxp3+ regulatory T cells. Adenosine production by these cells has been linked to the inhibition of CD4 T cell effector functions such as proliferation and cytokine secretion.
Description & Specifications

Specifications

CD73
Murine
Flow Cytometry
PerCP-eFluor 710
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.06μg)/test
AD2
Ecto-5'-nucleotidase, NT5E
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer