Please login to your online account to display your discounted pricing

CD73 Mouse anti-Human, PE, Clone: AD2, eBioscience™

Mouse Monoclonal Antibody

$158.05 - $390.05

Specifications

Antigen CD73
Clone AD2
Host Species Mouse
Gene Alias Ecto-5'-nucleotidase, NT5E
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50245275

 
life technologies
LS12073941
25 tests Each for $158.05

50245276

 
life technologies
LS12073942
100 tests Each for $390.05
Description & Specifications

Specifications

Antigen CD73
Clone AD2
Host Species Mouse
Gene Alias Ecto-5'-nucleotidase, NT5E
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

This AD2 monoclonal antibody reacts with human CD73, a 5’-ectonucleotidase that converts 5’-adenosine monophosphate to adenosine. CD73 is expressed on the surface of endothelial cells, as well as B and T cells, including some CD4+Foxp3+ regulatory T cells. Adenosine production by these cells has been linked to the inhibition of CD4 T cell effector functions such as proliferation and cytokine secretion.