missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD71 (Transferrin Receptor) Rat anti-Mouse, FITC, Clone: R17217 (RI7 217.1.4), eBioscience™

Rat Monoclonal Antibody

$118.80 - $381.48

Specifications

Antigen CD71 (Transferrin Receptor)
Clone R17217 (RI7 217.1.4)
Host Species Rat
Gene Alias TFRC
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
509657 Life Technologies
LS11071181
50μg Each for $118.80
509658 Life Technologies
LS11071182
100μg Each for $196.68
509659 Life Technologies
LS11071185
500μg Each for $381.48
Specifications

Description

The R17217 monoclonal antibody reacts with mouse CD71, a 170-180kDa type II transmembrane protein. CD71, the transferrin receptor, exists as a homodimer on the cell surface and is essential for cellular growth. CD71 is expressed by immature proliferating cells and at low levels on resting mature lymphocytes. Antigen or mitogen stimulation of T and B cells upregulates the expression of CD71. Expression level differences have been observed in different mouse strains.
Description & Specifications

Specifications

CD71 (Transferrin Receptor)
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
R17217 (RI7 217.1.4)
TFRC
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.5mg/mL