Please login to your online account to display your discounted pricing

CD71 (Transferrin Receptor) Rat anti-Mouse, Clone: R17217 (RI7 217.1.4), eBioscience™

Rat Monoclonal Antibody

$172.55 - $361.05

Specifications

Antigen CD71 (Transferrin Receptor)
Clone R17217 (RI7 217.1.4)
Host Species Rat
Gene Alias TFRC
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5012695

 
life technologies
LS14071182
100μg Each for $172.55

5012696

 
life technologies
LS14071185
500μg Each for $361.05
Description & Specifications

Specifications

Antigen CD71 (Transferrin Receptor)
Clone R17217 (RI7 217.1.4)
Host Species Rat
Gene Alias TFRC
Species Reactivity Mouse
Applications Flow Cytometry
Applications Functional Assay
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The R17217 monoclonal antibody reacts with mouse CD71, a 170-180kDa type II transmembrane protein. CD71, the transferrin receptor, exists as a homodimer on the cell surface and is essential for cellular growth. CD71 is expressed by immature proliferating cells and at low levels on resting mature lymphocytes. Antigen or mitogen stimulation of T and B cells upregulates the expression of CD71. Expression level differences have been observed in different mouse strains.