missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD71 (Transferrin Receptor) Mouse anti-Human, PE, Clone: OKT9 (OKT-9), eBioscience™

Mouse Monoclonal Antibody

$143.88 - $303.60

Specifications

Antigen CD71 (Transferrin Receptor)
Clone OKT9 (OKT-9)
Host Species Murine
Gene Alias TFRC
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5010469 Life Technologies
LS12071941
25 Tests Each for $143.88
5010470 Life Technologies
LS12071942
100 Tests Each for $303.60
Specifications

Description

The OKT9 monoclonal antibody reacts with human CD71, a 170-180kDa type II transmembrane protein. CD71, the transferrin receptor, exists as a homodimer on the cell surface and is essential for cellular growth. CD71 is expressed by immature proliferating cells and at low levels on resting mature lymphocytes. Antigen or mitogen stimulation of T and B cells upregulates the expression of CD71.
Description & Specifications

Specifications

CD71 (Transferrin Receptor)
Murine
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.06μg)/test
OKT9 (OKT-9)
TFRC
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer