Please login to your online account to display your discounted pricing

CD71 (Transferrin Receptor) Mouse anti-Human, Biotin, Clone: OKT9 (OKT-9), eBioscience™

Mouse Monoclonal Antibody

$114.55 - $259.55

Specifications

Antigen CD71 (Transferrin Receptor)
Clone OKT9 (OKT-9)
Host Species Mouse
Gene Alias TFRC
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011624

 
life technologies
LS13071980
25μg Each for $114.55

5011625

 
life technologies
LS13071982
100μg Each for $259.55
Description & Specifications

Specifications

Antigen CD71 (Transferrin Receptor)
Clone OKT9 (OKT-9)
Host Species Mouse
Gene Alias TFRC
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The OKT9 monoclonal antibody reacts with human CD71, a 170-180kDa type II transmembrane protein. CD71, the transferrin receptor, exists as a homodimer on the cell surface and is essential for cellular growth. CD71 is expressed by immature proliferating cells and at low levels on resting mature lymphocytes. Antigen or mitogen stimulation of T and B cells upregulates the expression of CD71.