Please login to your online account to display your discounted pricing

CD71 (Transferrin Receptor) Mouse anti-Human, APC, Clone: OKT9, eBioscience™

Mouse Monoclonal Antibody

$129.05 - $275.50

Specifications

Antigen CD71 (Transferrin Receptor)
Clone OKT9 (OKT-9)
Host Species Murine
Gene Alias TFRC
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5015020

 
life technologies
LS17071941
25 Tests Each for $129.05

5015021

 
life technologies
LS17071942
100 Tests Each for $275.50
Description & Specifications

Specifications

Antigen CD71 (Transferrin Receptor)
Clone OKT9 (OKT-9)
Host Species Murine
Gene Alias TFRC
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Allophycocyanin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.06μg)/test

The OKT9 monoclonal antibody reacts with human CD71, a 170-180kDa type II transmembrane protein. CD71, the transferrin receptor, exists as a homodimer on the cell surface and is essential for cellular growth. CD71 is expressed by immature proliferating cells and at low levels on resting mature lymphocytes. Antigen or mitogen stimulation of T and B cells upregulates the expression of CD71.