Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD71/Transferrin Receptor Ab-3, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry applications with Thermo Scientific™ CD71/Transferrin Receptor Ab-3, Mouse Monoclonal Antibody.

$274.80 - $1,023.65

Specifications

Antigen CD71/Transferrin Receptor Ab-3
Clone 10F11
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MS1096R7

 
thermo scientific
MS1096R7
7mL Each for $974.55

MS1096S

 
thermo scientific™ lab vision™
MS1096S
1mL Each for $1,023.65

MS1096S0

 
thermo scientific™ lab vision™
MS1096S0
100µL Each for $274.80

MS1096S1

 
thermo scientific™ lab vision™
MS1096S1
500µL Each for $679.80
Description & Specifications

Specifications

Antigen CD71/Transferrin Receptor Ab-3
Clone 10F11
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Mouse
Regulatory Status RUO
Type Primary
Isotype IgG2b

Ensure accurate, reproducible results in immunohistochemistry applications with Thermo Scientific™ CD71/Transferrin Receptor Ab-3, Mouse Monoclonal Antibody.

Cell surface transferrin receptor (CD71) is a disulfide-bonded homodimeric glycoprotein. Ligand for transferrin receptor is the serum iron transport protein, transferrin. This receptor is broadly distributed in carcinomas, sarcomas, leukemias, and lymphomas. Transferrin receptor (CD71) has been reported to be associated with cell proliferation in both normal and neoplastic tissues and useful in predicting clinical behavior or response to therapy in a number of malignancies including breast cancer.

Host Species: Mouse

Clone: 10F11

Isotype: IgG2b

Species Reactivity: Human. Others not known.

Epitope: Not determined

Molecular Weight: ∽90-95 kDa

Positive Control: Placenta

Cellular Localization: Granular cytoplasmic and membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)

Note:

USA: RUO; Int'l: RUO

General References:

1. Habeshaw J A, et al. Lancet. 1:498-500 (1983)

2. Lioyd J M, et al. Journal of Clinical Pathology. 37: 131-135 (1984).