missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD70 Rat anti-Mouse, PE, Clone: FR70, eBioscience™

Rat Monoclonal Antibody

$172.55 - $419.05

Specifications

Antigen CD70
Clone FR70
Host Species Rat
Gene Alias TNFSF7, Ki-24, CD27 ligand
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5010463 Life Technologies
LS12070181
50μg Each for $172.55
5010464 Life Technologies
LS12070182
100μg Each for $274.05
5010465 Life Technologies
LS12070183
200μg Each for $419.05
Specifications

Description

The FR70 monoclonal antibody reacts with mouse CD70, a 30-33kDa type II transmembrane glycoprotein. CD70 is a member of the TNF family and is expressed by activated mouse T and B lymphocytes. Interaction of CD70 with CD27, its ligand on T cells, is important in T-B cell crosstalk and plays a role in costimulation and immune regulation.
Description & Specifications

Specifications

CD70
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
FR70
TNFSF7, Ki-24, CD27 ligand
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL