missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD70 Rat anti-Mouse, Clone: FR70, eBioscience™

Rat Monoclonal Antibody

$64.68 - $328.68

Specifications

Antigen CD70
Clone FR70
Host Species Rat
Gene Alias TNFSF7, Ki-24, CD27 ligand
Applications Flow Cytometry,Functional Assay,Immunoprecipitation
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012692 Life Technologies
LS14070181
50μg Each for $64.68
5012693 Life Technologies
LS14070182
100μg Each for $130.68
5012694 Life Technologies
LS14070185
500μg Each for $328.68
Specifications

Description

The FR70 monoclonal antibody reacts with mouse CD70, a 30-33kDa type II transmembrane glycoprotein. CD70 is a member of the TNF family and is expressed by activated mouse T and B lymphocytes. Interaction of CD70 with CD27, its ligand on T cells, is important in T-B cell crosstalk and plays a role in costimulation and immune regulation.
Description & Specifications

Specifications

CD70
Rat
Flow Cytometry,Functional Assay,Immunoprecipitation
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL
FR70
TNFSF7, Ki-24, CD27 ligand
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer