missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD7 Mouse anti-Human, PerCP-eFluor 710, Clone: 4H9, eBioscience™

Mouse Monoclonal Antibody

$223.08 - $460.68

Specifications

Antigen CD7
Clone 4H9
Host Species Murine
Gene Alias Leu-9
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5015905 Life Technologies
LS46007841
25 Tests Each for $223.08
5015906 Life Technologies
LS46007842
100 Tests Each for $460.68
Specifications

Description

This 4H9 monoclonal antibody reacts with human CD7 (also known as Leu-9 or gp40), a 40kDa transmembrane glycoprotein belonging to the immunoglobulin superfamily. This molecule is expressed on peripheral blood T lymphocytes and natural killer cells, as well as on lymphoid and myeloid lineage precursors during adult hematopoiesis. Capable of providing a costimulatory signal and associating with CD3 and CD45, CD7 is involved in T cell activation.

CD7 has also been detected on numerous T cell-derived leukemias and lymphomias. In fact, because CD7 expression is greater on CD4+ T cells in classical Hodgkin’s lymphoma than in benign reactive lymph adenopathy, this molecule may be used as a diagnostic for Hodgkin’s lymphoma.

Blocking assays demonstrate that this clone recognizes the same epitope as clone 124-1D1.
Description & Specifications

Specifications

CD7
Murine
Flow Cytometry
PerCP-eFluor 710
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.25μg)/test
4H9
Leu-9
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer