missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD7 Mouse anti-Human, FITC, Clone: 4H9, eBioscience™

Mouse Monoclonal Antibody

$143.55 - $317.55

Specifications

Antigen CD7
Clone 4H9
Host Species Murine
Gene Alias Leu-9
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
509404 Life Technologies
LS11007841
25 Tests Each for $143.55
509405 Life Technologies
LS11007842
100 Tests Each for $317.55
Specifications

Description

This 4H9 monoclonal antibody reacts with human CD7 (also known as Leu-9 or gp40), a 40kDa transmembrane glycoprotein belonging to the immunoglobulin superfamily. This molecule is expressed on peripheral blood T lymphocytes and natural killer cells, as well as on lymphoid and myeloid lineage precursors during adult hematopoiesis. Capable of providing a costimulatory signal and associating with CD3 and CD45, CD7 is involved in T cell activation.

CD7 has also been detected on numerous T cell-derived leukemias and lymphomias. In fact, because CD7 expression is greater on CD4+ T cells in classical Hodgkin’s lymphoma than in benign reactive lymph adenopathy, this molecule may be used as a diagnostic for Hodgkin’s lymphoma.

Blocking assays demonstrate that this clone recognizes the same epitope as clone 124-1D1.
Description & Specifications

Specifications

CD7
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
4H9
Leu-9
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.125μg)/test