Please login to your online account to display your discounted pricing

CD69 Mouse anti-Human, FITC, Clone: FN50, eBioscience™

Mouse Monoclonal Antibody

$114.55 - $259.55

Specifications

Antigen CD69
Clone FN50
Host Species Mouse
Gene Alias Very Early Activation Antigen
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

509655

 
life technologies
LS11069941
25 tests Each for $114.55

501129436

 
life technologies
LS11069942
100 tests Each for $259.55
Description & Specifications

Specifications

Antigen CD69
Clone FN50
Host Species Mouse
Gene Alias Very Early Activation Antigen
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

The FN50 monoclonal antibody reacts with human CD69, also known as very early activation antigen (VEA). CD69 is approximately 30kDa and is expressed on the cell-surface as a disulfide-linked dimer. CD69 is rapidly upregulated upon activation and expressed on lymphocytes, monocytes and platelets.