missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD69 Mouse anti-Human, Clone: FN50, eBioscience™

Mouse Monoclonal Antibody

$64.68 - $130.68

Specifications

Antigen CD69
Clone FN50
Host Species Murine
Gene Alias Very Early Activation Antigen, VEA
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012690 Life Technologies
LS14069980
25μg Each for $64.68
5012691 Life Technologies
LS14069982
100μg Each for $130.68
Specifications

Description

The FN50 monoclonal antibody reacts with human CD69, also known as very early activation antigen (VEA). CD69 is approximately 30kDa and is expressed on the cell-surface as a disulfide-linked dimer. CD69 is rapidly upregulated upon activation and expressed on lymphocytes, monocytes and platelets.
Description & Specifications

Specifications

CD69
Murine
Human
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
FN50
Very Early Activation Antigen, VEA
Flow Cytometry,Immunohistochemistry
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL