missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD69 Mouse anti-Human, Biotin, Clone: FN50, eBioscience™

Mouse Monoclonal Antibody

$114.55 - $259.55

Specifications

Antigen CD69
Clone FN50
Host Species Murine
Gene Alias Very Early Activation Antigen, VEA
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122515 Life Technologies
LS13069982
100μg Each for $259.55
5011616 Life Technologies
LS13069980
25μg Each for $114.55
Specifications

Description

The FN50 monoclonal antibody reacts with human CD69, also known as very early activation antigen (VEA). CD69 is approximately 30kDa and is expressed on the cell-surface as a disulfide-linked dimer. CD69 is rapidly upregulated upon activation and expressed on lymphocytes, monocytes and platelets.
Description & Specifications

Specifications

CD69
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
FN50
Very Early Activation Antigen, VEA
Flow Cytometry
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL