missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD69 Mouse anti-Human, APC, Clone: FN50, eBioscience™

Mouse Monoclonal Antibody

$143.88 - $368.28

Specifications

Antigen CD69
Clone FN50
Host Species Murine
Gene Alias Very Early Activation Antigen, VEA
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5015016 Life Technologies
LS17069941
25 Tests Each for $143.88
5015017 Life Technologies
LS17069942
100 Tests Each for $368.28
Specifications

Description

The FN50 monoclonal antibody reacts with human CD69, also known as very early activation antigen (VEA). CD69 is approximately 30kDa and is expressed on the cell-surface as a disulfide-linked dimer. CD69 is rapidly upregulated upon activation and expressed on lymphocytes, monocytes and platelets.
Description & Specifications

Specifications

CD69
Murine
Flow Cytometry
Allophycocyanin
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.015μg)/test
FN50
Very Early Activation Antigen, VEA
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer