missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD69 Armenian Hamster anti-Mouse, PerCP-Cy5.5, Clone: H1.2F3, eBioscience™

Armenian Hamster Monoclonal Antibody

$143.88 - $381.48

Specifications

Antigen CD69
Clone H1.2F3
Host Species Armenian Hamster
Gene Alias Very Early Activation Antigen, VEA
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5015811 Life Technologies
LS45069182
100μg Each for $381.48
501130721 Life Technologies
LS45069180
25μg Each for $143.88
Specifications

Description

The H1.2F3 monoclonal antibody reacts with mouse CD69, also known as very early activation antigen (VEA). CD69 is approximately 35kDa and is expressed on the surface as a disulfide-linked dimer. While a small subset of lymphocytes in the thymus, spleen and lymph nodes express this antigen, activation of both T and B cells rapidly upregulates the surface expression of CD69, suggesting a role for CD69 in lymphocyte development and activation.
Description & Specifications

Specifications

CD69
Armenian Hamster
Flow Cytometry
PerCP-Cy5.5
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
H1.2F3
Very Early Activation Antigen, VEA
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer