Please login to your online account to display your discounted pricing

CD68 Mouse anti-Human, FITC, Clone: eBioY1/82A (Y1/82A), eBioscience™

Mouse Monoclonal Antibody

$145.00 - $294.35

Specifications

Antigen CD68
Clone eBioY1/82A (Y1/82A)
Host Species Mouse
Gene Alias macrosialin
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122077

 
life technologies
LS11068941
25 tests Each for $145.00

501122078

 
life technologies
LS11068942
100 tests Each for $294.35
Description & Specifications

Specifications

Antigen CD68
Clone eBioY1/82A (Y1/82A)
Host Species Mouse
Gene Alias macrosialin
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

The eBioY1/82 antibody reacts with CD68 which belongs to the sialomucin family and is closely related to the family of lysosomal-associated membrane proteins (lamps) and scavenger receptor. CD68 is predominantly an intracellular protein, found mainly in the late endosomal compartment but can be detected in smaller amounts on the surface of cells of mainly myeloid derived cells; monocytes and macrophages, dendritic cells (and in some langerhan cells), neutrophils, basophils mast cells, myeloid progenitor cells, and a subset of CD34+ hemopoietic bone marrow progenitor cells. The function has not been fully elucidated but based on it's homolgy and structure suggests a role in antigen processing or presentation and protection of the lysosomal membrane from hydrolytic enzymes. Reports have also seen expression in activated T cells, and about 40% of peripheral blood B-lymphocytes and 50% of all B-ALL.

Staining with eBioY1/82A shows surface staining on B cells and a minor percentage of monocytes while intracellular staining results in about 50% of B cells and most monocytes.