Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD68/Macrophage Marker Ab-3, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ CD68/Macrophage Marker Ab-3, Mouse Monoclonal Antibody.

$261.55 - $862.02

Specifications

Antigen CD68/Macrophage Marker Ab-3
Clone KP1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS397P

 
thermo scientific™ lab vision™
MS397P
Purified Ab with BSA and Azide 1mL Each for $792.35

MS397P0

 
thermo scientific™ lab vision™
MS397P0
Purified Ab with BSA and Azide 100µL Each for $261.55

MS397P1

 
thermo scientific™ lab vision™
MS397P1
Purified Ab with BSA and Azide 500µL Each for $626.22

MS397P1ABX

 
thermo scientific™ lab vision™
MS397P1ABX
Ab without BSA/Azide (Frozen) 100µL Each for $680.63

MS397PABX

 
thermo scientific™ lab vision™
MS397PABX
Ab without BSA/Azide (Frozen) 200µL Each for $862.02

MS397R7

 
thermo scientific™ lab vision™
MS397R7
Ready-to-use for IHC (UVLP) 7mL Each for $423.65
Description & Specifications

Specifications

Antigen CD68/Macrophage Marker Ab-3
Clone KP1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Host Species Mouse
Regulatory Status IVD
Research Discipline Hematopoietic
Type Primary
Immunogen Subcellular fraction of human alveolar macrophages

Obtain accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ CD68/Macrophage Marker Ab-3, Mouse Monoclonal Antibody.

CD68 is expressed on macrophages and monocytes.

Host Species: Mouse

Clone: KP1

Isotype: IgG1/κ

Species Reactivity: Human, Monkey, Cat and Rat. Does not react with pig, dog and chicken.

Epitope: Not determined

Immunogen: Subcellular fraction of human alveolar macrophages

Molecular Weight: 110 kDa

Positive Control: Tonsil, lymph node, or spleen

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)