Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD68/Macrophage Marker Ab-3, Mouse Monoclonal Antibody


Obtain accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ CD68/Macrophage Marker Ab-3, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS397P1ABX

 View more versions of this product

Catalog No. MS397P1ABX

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Non-Human Primate
Cross Reactivity Rat
Antigen CD68/Macrophage Marker Ab-3
Regulatory Status IVD
Host Species Mouse
Format Concentrated
Isotype IgG1, κ
Clone KP1
Quantity 100µL
Immunogen Subcellular fraction of human alveolar macrophages
Applications Immunohistochemistry (Paraffin)
Conjugate Unlabeled
Type Primary
Concentration 1mg/mL
Formulation Ab without BSA/Azide (Frozen)
Classification Monoclonal
Research Discipline Hematopoietic

Obtain accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ CD68/Macrophage Marker Ab-3, Mouse Monoclonal Antibody.

CD68 is expressed on macrophages and monocytes.

Host Species: Mouse

Clone: KP1

Isotype: IgG1/κ

Species Reactivity: Human, Monkey, Cat and Rat. Does not react with pig, dog and chicken.

Epitope: Not determined

Immunogen: Subcellular fraction of human alveolar macrophages

Molecular Weight: 110 kDa

Positive Control: Tonsil, lymph node, or spleen

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)