missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD66e (CEA) Mouse anti-Human, Clone: CB30, eBioscience™

Mouse Monoclonal Antibody

$130.68 - $341.88

Specifications

Antigen CD66e (CEA)
Clone CB30
Host Species Murine
Gene Alias Carcinoembryonic Antigen, CEACAM5
Applications Flow Cytometry,Immunohistochemistry,Western Blot
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012683 Life Technologies
LS14066980
25μg Each for $130.68
5012684 Life Technologies
LS14066982
100μg Each for $341.88
Specifications

Description

The monoclonal antibody CB30 recognizes human CD66e, also known as CEA (carcinoembryonic antigen) and CEACAM5. As a member of the immunoglobulin superfamily, it is large GPI-linked glycosylated protein with a molecular weight about 200 kDa. It is expressed mainly on epithelial cells and on a variety of tumors of the gastrointestinal, respiratory and genitourinary tracts. CD66e has been shown to function in cell adhesion and migration via homophilic and heterophilic interactions with NCA (non-specific cross-reacting antigen also known as CEACAM6 and CD66c). Overexpression is correlated with tumor invasiveness and is therefore used as a clinical marker.
Description & Specifications

Specifications

CD66e (CEA)
Murine
Flow Cytometry,Immunohistochemistry,Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL
CB30
Carcinoembryonic Antigen, CEACAM5
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer