Please login to your online account to display your discounted pricing

CD66b Mouse anti-Human, PE-Cy7, Clone: G10F5, eBioscience™

Mouse Monoclonal Antibody

$191.40 - $471.25

Specifications

Antigen CD66b
Clone G10F5
Host Species Murine
Gene Alias CGM6, carcinoembryonic antigen gene family member 6
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123300

 
life technologies
LS25066641
25 Tests Each for $191.40

501129222

 
life technologies
LS25066642
100 Tests Each for $471.25
Description & Specifications

Specifications

Antigen CD66b
Clone G10F5
Host Species Murine
Gene Alias CGM6, carcinoembryonic antigen gene family member 6
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy7
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.06μg)/test

This G10F5 monoclonal antibody reacts with human CD66b, one of four members of the CD66 family of granulocyte-specific activation antigens. Expressed on neutrophils and eosinophils, CD66b is upregulated following stimulation. This glycosyl-phosphatidylinositol-linked cell surface receptor plays a role in regulating neutrophil activation and adhesion to fibronectin, galectin-3, and E-selectin.