missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD66b Mouse anti-Human, PE, Clone: G10F5, eBioscience™

Mouse Monoclonal Antibody

$162.40 - $397.30

Specifications

Antigen CD66b
Clone G10F5
Host Species Murine
Gene Alias CGM6, carcinoembryonic antigen gene family member 6
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122245 Life Technologies
LS12066641
25 Tests Each for $162.40
501122246 Life Technologies
LS12066642
100 Tests Each for $397.30
Specifications

Description

This G10F5 monoclonal antibody reacts with human CD66b, one of four members of the CD66 family of granulocyte-specific activation antigens. Expressed on neutrophils and eosinophils, CD66b is upregulated following stimulation. This glycosyl-phosphatidylinositol-linked cell surface receptor plays a role in regulating neutrophil activation and adhesion to fibronectin, galectin-3, and E-selectin.
Description & Specifications

Specifications

CD66b
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
G10F5
CGM6, carcinoembryonic antigen gene family member 6
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.125μg)/test