missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD66b Mouse anti-Human, FITC, Clone: G10F5, eBioscience™

Mouse Monoclonal Antibody

$132.00 - $266.64

Specifications

Antigen CD66b
Clone G10F5
Host Species Murine
Gene Alias CGM6, carcinoembryonic antigen gene family member 6
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122075 Life Technologies
LS11066641
25 Tests Each for $132.00
501122076 Life Technologies
LS11066642
100 Tests Each for $266.64
Specifications

Description

This G10F5 monoclonal antibody reacts with human CD66b, one of four members of the CD66 family of granulocyte-specific activation antigens. Expressed on neutrophils and eosinophils, CD66b is upregulated following stimulation. This glycosyl-phosphatidylinositol-linked cell surface receptor plays a role in regulating neutrophil activation and adhesion to fibronectin, galectin-3, and E-selectin.
Description & Specifications

Specifications

CD66b
Murine
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.5μg)/test
G10F5
CGM6, carcinoembryonic antigen gene family member 6
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer