missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD64 (Fc gamma Receptor 1) Mouse anti-Human, APC-eFluor 780, Clone: 10.1, eBioscience™

Mouse Monoclonal Antibody

$172.55 - $442.25

Specifications

Antigen CD64 (Fc gamma Receptor 1)
Clone 10.1
Host Species Murine
Gene Alias FcgR1
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124035 Life Technologies
LS47064941
25 Tests Each for $172.55
501124036 Life Technologies
LS47064942
100 Tests Each for $442.25
Specifications

Description

The 10.1 monoclonal antibody reacts with human CD64 (FcRI), a 75kDa type I transmembrane protein. CD64 is the high affinity receptor for IgG and is involved in antibody-dependent cell-mediated cytotoxicity (ADCC), phagocytosis, and regulation of cytokine production. Monocytes and macrophages express CD64, while mature granulocytes and lymphocytes are negative.
Description & Specifications

Specifications

CD64 (Fc gamma Receptor 1)
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
10.1
FcgR1
Flow Cytometry
APC-eFluor 780
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
5μL (0.125μg)/test