Please login to your online account to display your discounted pricing

CD63 Rat anti-Mouse, PE-Cy7, Clone: NVG-2, eBioscience™

Rat Monoclonal Antibody

$201.55 - $433.55

Specifications

Antigen CD63
Clone NVG-2
Host Species Rat
Gene Alias LAMP3, DC-LAMP, LIMP-1, TSPAN30, MLA1, ME491, granulophysin
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123296

 
life technologies
LS25063180
25μg Each for $201.55

501123297

 
life technologies
LS25063182
100μg Each for $433.55
Description & Specifications

Specifications

Antigen CD63
Clone NVG-2
Host Species Rat
Gene Alias LAMP3, DC-LAMP, LIMP-1, TSPAN30, MLA1, ME491, granulophysin
Species Reactivity Mouse
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate PE-Cy7
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The monoclonal antibody NVG-2 reacts with mouse CD63, also known as Lysosomal-Associated Membrane Protein 3 (LAMP-3) or tetraspanin 30 (TSPN30), a member of tetraspanin family of proteins characterized by four transmembrane domains. CD63 is expressed on a variety of cell types of hematopoietic lineage, e.g., granulocytes, B lymphocytes, platelets, as well as cells of non-hematopoietic origin. It can be found on the cell membrane, late endocytic vesicles, lysosomes, exosomes, and other specialized granules. On the cell surface, CD63 has been shown to interact with various proteins forming tetraspanin-enriched microdomains (TEM). Its high expression on the cell membrane may be indicative of cell activation, hence, CD63 is often used as an activation marker for basophils, platelets and other cells.