missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD63 Mouse anti-Human, PE-Cy7, Clone: H5C6, eBioscience™

Mouse Monoclonal Antibody

$178.35 - $456.75

Specifications

Antigen CD63
Clone H5C6
Host Species Murine
Gene Alias Lysosome-associated membrane protein 3, Lamp 3
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123298 Life Technologies
LS25063941
25 Tests Each for $178.35
501123299 Life Technologies
LS25063942
100 Tests Each for $456.75
Specifications

Description

This H5C6 monoclonal antibody reacts with human CD63, a type III member of the tetraspanin family of transmembrane proteins. CD63 is expressed intracellularly on lysosomes, endosomes, and granules of resting platelets and basophils. However, cell surface expression of CD63 can be detected on activated basophils and platelets, monocytes, macrophages, and granulocytes. This receptor is also expressed on endothelial cells, fibroblasts, and smooth muscle cells. Studies have demonstrated that CD63 associates with integrins (VLA-3 and VLA-6) and TIMP-1 to mediate the allergic response.
Description & Specifications

Specifications

CD63
Murine
Flow Cytometry
PE-Cy7
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
5μL (0.5μg)/test
H5C6
Lysosome-associated membrane protein 3, Lamp 3
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer