Please login to your online account to display your discounted pricing

CD63 Mouse anti-Human, PE, Clone: H5C6, eBioscience™

Mouse Monoclonal Antibody

$152.25 - $375.55

Specifications

Antigen CD63
Clone H5C6
Host Species Mouse
Gene Alias Lysosome-associated membrane protein 3, Lamp 3
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122243

 
life technologies
LS12063941
25 tests Each for $152.25

501122244

 
life technologies
LS12063942
100 tests Each for $375.55
Description & Specifications

Specifications

Antigen CD63
Clone H5C6
Host Species Mouse
Gene Alias Lysosome-associated membrane protein 3, Lamp 3
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

This H5C6 monoclonal antibody reacts with human CD63, a type III member of the tetraspanin family of transmembrane proteins. CD63 is expressed intracellularly on lysosomes, endosomes, and granules of resting platelets and basophils. However, cell surface expression of CD63 can be detected on activated basophils and platelets, monocytes, macrophages, and granulocytes. This receptor is also expressed on endothelial cells, fibroblasts, and smooth muscle cells. Studies have demonstrated that CD63 associates with integrins (VLA-3 and VLA-6) and TIMP-1 to mediate the allergic response.