missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD62P (P-Selectin) Mouse anti-Human, PE, Clone: AK-4, eBioscience™

Mouse Monoclonal Antibody

$130.68 - $289.08

Specifications

Antigen CD62P (P-Selectin)
Clone AK-4
Host Species Murine
Gene Alias LECAM-3
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5010448 Life Technologies
LS12062842
100 Tests Each for $289.08
5010447 Life Technologies
LS12062841
25 Tests Each for $130.68
Specifications

Description

The AK-4 monoclonal antibody reacts with human CD62P, a 140kDa transmembrane glycoprotein also known as P-Selectin. Upon activation, CD62P is rapidly transported from its cytoplasmic stores in the Weibel-Palade bodies of endothelial cells or the alpha-granules of platelets to the surface. CD62P initiates the adhesive interaction between endothelial cells and neutrophils and monocytes during the inflammatory reaction and is also involved in the interaction of platelets with monocytes and neutrophils.
Description & Specifications

Specifications

CD62P (P-Selectin)
Murine
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.125μg)/test
AK-4
LECAM-3
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer