Please login to your online account to display your discounted pricing

CD62P (P-Selectin) Mouse anti-Human, FITC, Clone: AK-4, eBioscience™

Mouse Monoclonal Antibody

$114.55 - $274.05

Specifications

Antigen CD62P (P-Selectin)
Clone AK-4
Host Species Murine
Gene Alias LECAM-3
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

509644

 
life technologies
LS11062841
25 Tests Each for $114.55

509645

 
life technologies
LS11062842
100 Tests Each for $274.05
Description & Specifications

Specifications

Antigen CD62P (P-Selectin)
Clone AK-4
Host Species Murine
Gene Alias LECAM-3
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (1μg)/test

The AK-4 monoclonal antibody reacts with human CD62P, a 140kDa transmembrane glycoprotein also known as P-Selectin. Upon activation, CD62P is rapidly transported from its cytoplasmic stores in the Weibel-Palade bodies of endothelial cells or the alpha-granules of platelets to the surface. CD62P initiates the adhesive interaction between endothelial cells and neutrophils and monocytes during the inflammatory reaction and is also involved in the interaction of platelets with monocytes and neutrophils.