missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD62P (P-Selectin) Mouse anti-Human, Clone: AK-4, eBioscience™

Mouse Monoclonal Antibody

$64.68 - $130.68

Specifications

Antigen CD62P (P-Selectin)
Clone AK-4
Host Species Murine
Gene Alias LECAM-3
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012675 Life Technologies
LS14062880
25μg Each for $64.68
5012676 Life Technologies
LS14062882
100μg Each for $130.68
Specifications

Description

The AK-4 monoclonal antibody reacts with human CD62P, a 140kDa transmembrane glycoprotein also known as P-Selectin. Upon activation, CD62P is rapidly transported from its cytoplasmic stores in the Weibel-Palade bodies of endothelial cells or the alpha-granules of platelets to the surface. CD62P initiates the adhesive interaction between endothelial cells and neutrophils and monocytes during the inflammatory reaction and is also involved in the interaction of platelets with monocytes and neutrophils.
Description & Specifications

Specifications

CD62P (P-Selectin)
Murine
Human
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
AK-4
LECAM-3
Flow Cytometry,Functional Assay,Immunohistochemistry (Frozen)
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL