Please login to your online account to display your discounted pricing

CD62L (L-selectin) Rat anti-Mouse, PE-Cy7, Clone: MEL-14, eBioscience™

Rat Monoclonal Antibody

$172.55 - $324.80

Specifications

Antigen CD62L (L-selectin)
Clone MEL-14
Host Species Rat
Gene Alias LECAM-1, Ly-22
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5015527

 
life technologies
LS25062181
50μg Each for $172.55

5015528

 
life technologies
LS25062182
100μg Each for $324.80
Description & Specifications

Specifications

Antigen CD62L (L-selectin)
Clone MEL-14
Host Species Rat
Gene Alias LECAM-1, Ly-22
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy7
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The MEL-14 monoclonal antibody reacts with mouse CD62L, a 76kDa member of the selectin family. CD62L is expressed by neutrophils, monocytes, and subsets of T, B, and NK cells and binds a number of glycosylated, fucosylated, sulfated sialylated glycoproteins including CD34, glycam-1 and MAdCam-1. These interactions mediate rolling of lymphocytes on activated endothelium at the sites of inflammation and homing of cells to the high endothelial venules (HEV) of peripheral lymphoid tissues.