Please login to your online account to display your discounted pricing

CD62L (L-selectin) Rat anti-Mouse, FITC, Clone: MEL-14, eBioscience™

Rat Monoclonal Antibody

$114.55 - $709.05

Specifications

Antigen CD62L (L-selectin)
Clone MEL-14
Host Species Rat
Gene Alias LECAM-1, Ly-22
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

509639

 
affymetrix
11062182
100μg Each for $201.55

509638

 
affymetrix
11062181
50μg Each for $114.55

509640

 
affymetrix
11062185
500μg Each for $419.05

509641

 
affymetrix
11062186
1mg Each for $709.05
Description & Specifications

Specifications

Antigen CD62L (L-selectin)
Clone MEL-14
Host Species Rat
Gene Alias LECAM-1, Ly-22
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The MEL-14 monoclonal antibody reacts with mouse CD62L, a 76kDa member of the selectin family. CD62L is expressed by neutrophils, monocytes, and subsets of T, B, and NK cells and binds a number of glycosylated, fucosylated, sulfated sialylated glycoproteins including CD34, glycam-1 and MAdCam-1. These interactions mediate rolling of lymphocytes on activated endothelium at the sites of inflammation and homing of cells to the high endothelial venules (HEV) of peripheral lymphoid tissues.