missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD62L (L-selectin) Rat anti-Mouse, APC-eFluor 780, Clone: MEL-14, eBioscience™

Rat Monoclonal Antibody

$145.00 - $353.80

Specifications

Antigen CD62L (L-selectin)
Clone MEL-14
Host Species Rat
Gene Alias LECAM-1, Ly-22
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5016176 Life Technologies
LS47062182
100μg Each for $353.80
501129560 Life Technologies
LS47062180
25μg Each for $145.00
Specifications

Description

The MEL-14 monoclonal antibody reacts with mouse CD62L, a 76kDa member of the selectin family. CD62L is expressed by neutrophils, monocytes, and subsets of T, B, and NK cells and binds a number of glycosylated, fucosylated, sulfated sialylated glycoproteins including CD34, glycam-1 and MAdCam-1. These interactions mediate rolling of lymphocytes on activated endothelium at the sites of inflammation and homing of cells to the high endothelial venules (HEV) of peripheral lymphoid tissues.
Description & Specifications

Specifications

CD62L (L-selectin)
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
MEL-14
LECAM-1, Ly-22
Flow Cytometry
APC-eFluor 780
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
0.2mg/mL