missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD62L (L-selectin) Mouse anti-Rat, eFluor 660, Clone: OX85, eBioscience™

Mouse Monoclonal Antibody

$130.68 - $328.68

Specifications

Antigen CD62L (L-selectin)
Clone OX85
Host Species Murine
Species Reactivity Rat
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124277 Life Technologies
LS50062380
25μg Each for $130.68
501124278 Life Technologies
LS50062382
100μg Each for $328.68
Specifications

Description

The OX85 monoclonal antibody reacts with rat CD62L, which is also known as L-Selectin. CD62L is a 76kDa member of the selectin family that is expressed by neutrophils, monocytes, and subsets of T, B, and NK cells. This cell surface receptor binds glycosylated, fucosylated, sulfated, and sialylated glycoproteins including CD34, GlyCAM-1, and MadCAM-1. These interactions mediate lymphocyte rolling on activated endothelium at sites of inflammation and homing to high endothelial venules of peripheral lymphoid tissues.
Description & Specifications

Specifications

CD62L (L-selectin)
Murine
Flow Cytometry
eFluor 660
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
OX85
Rat
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer