Please login to your online account to display your discounted pricing

CD62L (L-selectin) Mouse anti-Human, PE-Cy7, Clone: DREG-56 (DREG56), eBioscience™

Mouse Monoclonal Antibody

$216.05 - $433.55

Specifications

Antigen CD62L (L-selectin)
Clone DREG-56 (DREG56)
Host Species Mouse
Gene Alias LECAM-1, Leu-8
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5015529

 
affymetrix
25062941
25 tests Each for $216.05

501129238

 
affymetrix
25062942
100 tests Each for $433.55
Description & Specifications

Specifications

Antigen CD62L (L-selectin)
Clone DREG-56 (DREG56)
Host Species Mouse
Gene Alias LECAM-1, Leu-8
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy7
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.5μg)/test

The DREG-56 monoclonal antibody reacts with human CD62L, a 76kDa member of the selectin family. CD62L is expressed by neutrophils, monocytes, and subsets of T, B, and NK cells and binds a number of glycosylated, fucosylated, sulfated sialylated glycoproteins including CD34, glycam-1 and MAdCAM-1. These interactions mediate rolling of lymphocytes on activated endothelium at the sites of inflammation and homing of cells to the high endothelial venules (HEV) of peripheral lymphoid tissues.