missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD62L (L-selectin) Mouse anti-Human, PE-Cy5, Clone: DREG-56 (DREG56), eBioscience™

Mouse Monoclonal Antibody

$172.55 - $375.55

Specifications

Antigen CD62L (L-selectin)
Clone DREG-56 (DREG56)
Host Species Murine
Gene Alias LECAM-1, Leu-8
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5014070 Life Technologies
LS15062941
25 Tests Each for $172.55
5014071 Life Technologies
LS15062942
100 Tests Each for $375.55
Specifications

Description

The DREG-56 monoclonal antibody reacts with human CD62L, a 76kDa member of the selectin family. CD62L is expressed by neutrophils, monocytes, and subsets of T, B, and NK cells and binds a number of glycosylated, fucosylated, sulfated sialylated glycoproteins including CD34, glycam-1 and MAdCAM-1. These interactions mediate rolling of lymphocytes on activated endothelium at the sites of inflammation and homing of cells to the high endothelial venules (HEV) of peripheral lymphoid tissues.
Description & Specifications

Specifications

CD62L (L-selectin)
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
DREG-56 (DREG56)
LECAM-1, Leu-8
Flow Cytometry
PE-Cy5
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.5μg)/test