missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD62L (L-selectin) Mouse anti-Human, PE, Clone: DREG-56 (DREG56), eBioscience™

Mouse Monoclonal Antibody

$117.48 - $289.08

Specifications

Antigen CD62L (L-selectin)
Clone DREG-56 (DREG56)
Host Species Murine
Gene Alias LECAM-1, Leu-8
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5010449 Life Technologies
LS12062941
25 Tests Each for $117.48
5010450 Life Technologies
LS12062942
100 Tests Each for $289.08
Specifications

Description

The DREG-56 monoclonal antibody reacts with human CD62L, a 76kDa member of the selectin family. CD62L is expressed by neutrophils, monocytes, and subsets of T, B, and NK cells and binds a number of glycosylated, fucosylated, sulfated sialylated glycoproteins including CD34, glycam-1 and MAdCAM-1. These interactions mediate rolling of lymphocytes on activated endothelium at the sites of inflammation and homing of cells to the high endothelial venules (HEV) of peripheral lymphoid tissues.
Description & Specifications

Specifications

CD62L (L-selectin)
Murine
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.125μg)/test
DREG-56 (DREG56)
LECAM-1, Leu-8
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer