Please login to your online account to display your discounted pricing

CD62E (E-Selectin) Mouse anti-Human, PE, Clone: P2H3, eBioscience™

Mouse Monoclonal Antibody

$158.05 - $332.05

Specifications

Antigen CD62E (E-Selectin)
Clone P2H3
Host Species Murine
Gene Alias LECAM-2
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122241

 
life technologies
LS12062741
25 Tests Each for $158.05

501122242

 
life technologies
LS12062742
100 Tests Each for $332.05
Description & Specifications

Specifications

Antigen CD62E (E-Selectin)
Clone P2H3
Host Species Murine
Gene Alias LECAM-2
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.5μg)/test

The P2H3 monoclonal antibody reacts with human CD62E, a 97-115kDa member of the selectin family. CD62E, also known as E-selectin or endothelial-leukocyte adhesion molecule-1 (ELAM-1) is an adhesion molecule expressed by endothelial cells upon stimulation with cytokines including TNFα and IL-1β. Induced expression of CD62E during inflammatory conditions is thought to mediate leukocyte rolling including the initial interaction of neutrophils with endothelium.

The P2H3 monoclonal antibody also inhibits cellular adhesion to cytokine-activated endothelial cells.