missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD62E (E-Selectin) Mouse anti-Human, Clone: P2H3, eBioscience™

Mouse Monoclonal Antibody

$100.05 - $172.55

Specifications

Antigen CD62E (E-Selectin)
Clone P2H3
Host Species Murine
Gene Alias LECAM-2
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012673 Life Technologies
LS14062780
25μg Each for $100.05
5012674 Life Technologies
LS14062782
100μg Each for $172.55
Specifications

Description

The P2H3 monoclonal antibody reacts with human CD62E, a 97-115kDa member of the selectin family. CD62E, also known as E-selectin or endothelial-leukocyte adhesion molecule-1 (ELAM-1) is an adhesion molecule expressed by endothelial cells upon stimulation with cytokines including TNFα and IL-1β. Induced expression of CD62E during inflammatory conditions is thought to mediate leukocyte rolling including the initial interaction of neutrophils with endothelium.

The P2H3 monoclonal antibody also inhibits cellular adhesion to cytokine-activated endothelial cells.
Description & Specifications

Specifications

CD62E (E-Selectin)
Murine
Human
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
P2H3
LECAM-2
Flow Cytometry,Functional Assay,Immunohistochemistry,Immunoprecipitation
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL