missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD61 (Integrin beta 3) Mouse anti-Human, FITC, Clone: VI-PL2, eBioscience™

Mouse Monoclonal Antibody

$117.48 - $275.88

Specifications

Antigen CD61 (Integrin beta 3)
Clone VI-PL2
Host Species Murine
Gene Alias ITGB3
Species Reactivity Baboon, Canine, Cynomolgus, Human, Rhesus Ma
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122074 Life Technologies
LS11061941
25 Tests Each for $117.48
509637 Life Technologies
LS11061942
100 Tests Each for $275.88
Specifications

Description

The VI-PL2 monoclonal antibody reacts with human CD61, also known as integrin beta 3 and GPIIIa. CD61 is a 90-110kDa member of the beta integrin family expressed by a wide variety of cells, including leukocytes, platelets, endothelial and smooth muscle cells. CD61 binds non-covalently with the alpha integrins CD41 and CD51, to form the alpha IIb beta3 (CD41/CD61) and alpha v beta3 (CD51/CD61) complexes. These alpha beta heterodimers are capable of mediating a variety of cellular responses including adhesion, trafficking, proliferaton and differentiation. The CD41/CD61 integrin has been shown to be involved in platelet aggregation, and binds to fibrinogen, von Willebrand Factor (vWF) and fibronectin. The CD51/CD61 integrin binds to matrix proteins including vitronectin, fibronectin, vWF and fibrinogen, and has been shown to have a strong role in modulating the migration and survival of angiogenic endothelial cells.

This VI-PL2 antibody has also been shown to crossreact with canine and several non-human primate species, including baboon, rhesus, and cynomologus monkey.
Description & Specifications

Specifications

CD61 (Integrin beta 3)
Murine
Baboon, Canine, Cynomolgus, Human, Rhesus Ma
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
VI-PL2
ITGB3
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.25μg)/test