Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD61 (GPIIIa) Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ CD61 (GPIIIa) Ab-1, Mouse Monoclonal Antibody.

$348.16 - $919.37

Specifications

Antigen CD61 (GPIIIa) Ab-1
Clone 2f2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MS1095R7

 
thermo scientific™ lab vision™
MS1095R7
7mL Each for $757.27

MS1095S

 
thermo scientific™ lab vision™
MS1095S
1mL Each for $919.37

MS1095S0

 
thermo scientific™ lab vision™
MS1095S0
100µL Each for $348.16
Description & Specifications

Specifications

Antigen CD61 (GPIIIa) Ab-1
Clone 2f2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Mouse
Regulatory Status RUO
Research Discipline Hematopoietic
Type Primary
Immunogen Recombinant protein encoding part of the external domain of human CD61
Isotype IgG1

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ CD61 (GPIIIa) Ab-1, Mouse Monoclonal Antibody.

CD61 (GPIIIa) is a glycoprotein found on megakaryocytes, platelets and their precursors. CD61 antigen plays a role in platelet aggregation and also as a receptor for fibrinogen, fibronectin, von Willebrand factor and vitronectrin.

Host Species: Mouse

Clone: 2f2

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Extracellular domain

Immunogen: Recombinant protein encoding part of the external domain of human CD61

Molecular Weight: 105 kDa

Positive Control: Tonsil

Cellular Localization: Membrane of platelets

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO