missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD58 (LFA-3) Mouse anti-Human, APC, Clone: TS2/9, eBioscience™

Mouse Monoclonal Antibody

$132.00 - $335.28

Specifications

Antigen CD58 (LFA-3)
Clone TS2/9
Host Species Murine
Applications Flow Cytometry
Regulatory Status RUO
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5015003 Life Technologies
LS17057841
25 Tests Each for $132.00
5015004 Life Technologies
LS17057842
100 Tests Each for $335.28
Specifications

Description

The TS2/9 antibody recognizes human CD58, which is also known as Lymphocyte Associated Antigen-3 (LFA-3). CD58 exists both as a transmembrane protein as well as a GPI-linked extracellular protein. It has been shown that cells can express both forms concurrently. In addition, soluble CD58 has also been observed in serum, urine and synoval fluid of rheumatoid arthritis patients. CD58 is found on non-hematopoietic and most hematopoietic cells. Binding to its ligand CD2, which is present on T cells, initiates a signaling pathway which includes phosphorylation and results in release of cytokines. CD58 has been shown to be important in adhesion to various cell types, T cell activation, cytokine production and cytoxicity.
The TS2/9 antibody has been reported to induce signaling events upon crosslinking. The epitope is in domain 1 of the immunoglobulin region.
Description & Specifications

Specifications

CD58 (LFA-3)
Murine
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
TS2/9
Flow Cytometry
Allophycocyanin
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.5μg)/test