Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD57 (Natural Killer Cell Marker) Ab-2, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry and flow cytometry experiments.

$243.04 - $1,209.59

Specifications

Antigen CD57 (Natural Killer Cell Marker) Ab-2
Clone HNK-1; same as Leu-7
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Flow Cytometry
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS1163P

 
thermo scientific™ lab vision™
MS1163P
Purified Ab with BSA and Azide 1mL Each for $1,113.63

MS1163P0

 
thermo scientific™ lab vision™
MS1163P0
Purified Ab with BSA and Azide 100μL Each for $243.04

MS1163P1

 
thermo scientific™ lab vision™
MS1163P1
Purified Ab with BSA and Azide 500μL Each for $706.96

MS1163P1ABX

 
thermo scientific™ lab vision™
MS1163P1ABX
Ab without BSA/Azide (Frozen) 100μL Each for $792.65

MS1163PABX

 
thermo scientific™ lab vision™
MS1163PABX
Ab without BSA/Azide (Frozen) 200μL Each for $1,209.59

MS1163R7

 
thermo scientific
MS1163R7
Ready-to-use for IHC (UVLP) 7mL Each for $660.55
Description & Specifications

Specifications

Antigen CD57 (Natural Killer Cell Marker) Ab-2
Clone HNK-1; same as Leu-7
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Murine
Regulatory Status RUO
Research Discipline Cell Adhesion
Type Primary
Immunogen Membrane extracts of the HSB-2 T lymphoblastoid cell line
Isotype IgM kappa

Obtain accurate, reproducible results in immunohistochemistry and flow cytometry experiments with Thermo Scientific™ CD57 (Natural Killer Cell Marker) Ab-2, Mouse Monoclonal Antibody.

CD57/HNK-1 is expressed on a subpopulation of 15-20% of peripheral blood mononuclear cells, about 60% of NK active cells and on a subset of T cells.

Host Species: Mouse

Clone: HNK-1; same as Leu-7

Isotype: IgM/κ

Species Reactivity: Human. Others not known.

Immunogen: Membrane extracts of the HSB-2 T-lymphoblastoid cell line

Molecular Weight: 110 kDa

Positive Control: Lymph node or tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO