Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD57 (Natural Killer Cell Marker) Ab-2, Mouse Monoclonal Antibody


Obtain accurate, reproducible results in immunohistochemistry and flow cytometry experiments.

Manufacturer: thermo scientific™ lab vision™  MS1163PABX

 View more versions of this product

Catalog No. MS1163PABX

  • / Each
 

Description & Specifications

Specifications

Antigen CD57 (Natural Killer Cell Marker) Ab-2
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HNK-1; same as Leu-7
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Formulation Ab without BSA/Azide (Frozen)
Host Species Murine
Immunogen Membrane extracts of the HSB-2 T lymphoblastoid cell line
Isotype IgM kappa
Quantity 200μL
Regulatory Status RUO
Research Discipline Cell Adhesion
Type Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human

Obtain accurate, reproducible results in immunohistochemistry and flow cytometry experiments with Thermo Scientific™ CD57 (Natural Killer Cell Marker) Ab-2, Mouse Monoclonal Antibody.

CD57/HNK-1 is expressed on a subpopulation of 15-20% of peripheral blood mononuclear cells, about 60% of NK active cells and on a subset of T cells.

Host Species: Mouse

Clone: HNK-1; same as Leu-7

Isotype: IgM/κ

Species Reactivity: Human. Others not known.

Immunogen: Membrane extracts of the HSB-2 T-lymphoblastoid cell line

Molecular Weight: 110 kDa

Positive Control: Lymph node or tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO