Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD57 (Natural Killer Cell Marker) Ab-2, Mouse Monoclonal Antibody


Obtain accurate, reproducible results in immunohistochemistry and flow cytometry experiments.

Manufacturer: thermo scientific™ lab vision™  MS1163P1ABX

 View more versions of this product

Catalog No. MS1163P1ABX

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Quantity 100µL
Cross Reactivity Human
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Immunogen Membrane extracts of the HSB-2 T lymphoblastoid cell line
Regulatory Status RUO
Clone HNK-1; same as Leu-7
Antigen CD57 (Natural Killer Cell Marker) Ab-2
Format Concentrated
Isotype IgM kappa
Formulation Ab without BSA/Azide (Frozen)
Research Discipline Cell Adhesion
Classification Monoclonal
Type Primary
Concentration 1mg/mL
Conjugate Unlabeled
Item Description 100uL; 1mg/mL; Purified w/o BSA, azide

Obtain accurate, reproducible results in immunohistochemistry and flow cytometry experiments with Thermo Scientific™ CD57 (Natural Killer Cell Marker) Ab-2, Mouse Monoclonal Antibody.

CD57/HNK-1 is expressed on a subpopulation of 15-20% of peripheral blood mononuclear cells, about 60% of NK active cells and on a subset of T cells.

Host Species: Mouse

Clone: HNK-1; same as Leu-7

Isotype: IgM/κ

Species Reactivity: Human. Others not known.

Immunogen: Membrane extracts of the HSB-2 T-lymphoblastoid cell line

Molecular Weight: 110 kDa

Positive Control: Lymph node or tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO