Please login to your online account to display your discounted pricing

CD57 Mouse anti-Human, PE-Cy7, Clone: TB01 (TBO1), eBioscience™

Mouse Monoclonal Antibody

$216.05 - $442.25

Specifications

Antigen CD57
Clone TB01 (TBO1)
Host Species Mouse
Gene Alias Leu7, NK1
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123294

 
life technologies
LS25057741
25 tests Each for $216.05

501123295

 
life technologies
LS25057742
100 tests Each for $442.25
Description & Specifications

Specifications

Antigen CD57
Clone TB01 (TBO1)
Host Species Mouse
Gene Alias Leu7, NK1
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy7
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (1.0μg)/test

This TB01 monoclonal antibody reacts with human CD57 (also known as HNK-1 and Leu-7), a 110kDa cell surface glycoprotein expressed on a subset of natural killer (NK) cells and NK T cells.