missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD57 Mouse anti-Human, FITC, Clone: TB01 (TBO1), eBioscience™

Mouse Monoclonal Antibody

$143.55 - $290.00

Specifications

Antigen CD57
Clone TB01 (TBO1)
Host Species Murine
Gene Alias Leu7, NK1
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
509627 Life Technologies
LS11057741
25 Tests Each for $143.55
509628 Life Technologies
LS11057742
100 Tests Each for $290.00
Specifications

Description

This TB01 monoclonal antibody reacts with human CD57 (also known as HNK-1 and Leu-7), a 110kDa cell surface glycoprotein expressed on a subset of natural killer (NK) cells and NK T cells.
Description & Specifications

Specifications

CD57
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
TB01 (TBO1)
Leu7, NK1
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (1.0μg)/test