Please login to your online account to display your discounted pricing

CD56 (NCAM) Mouse anti-Human, PE-eFluor 610, Clone: CMSSB, eBioscience™

Mouse Monoclonal Antibody

$187.05 - $448.05

Specifications

Antigen CD56 (NCAM)
Clone CMSSB
Host Species Mouse
Gene Alias Neural cell adhesion molecule
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124768

 
life technologies
LS61056741
25 tests Each for $187.05

501124769

 
life technologies
LS61056742
100 tests Each for $448.05
Description & Specifications

Specifications

Antigen CD56 (NCAM)
Clone CMSSB
Host Species Mouse
Gene Alias Neural cell adhesion molecule
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-eFluor 610
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

This CMSSB monoclonal antibody reacts with human CD56, also known as Neural Cell Adhesion Molecule (NCAM). CD56 is a highly glycosylated transmembrane molecule expressed by neurons and plays a role in the homotypic adhesion of neural cells. In the hematopoietic system, CD56 is expressed on NK cells and a subset of T cells referred to as NKT cells. Staining with CMSSB does not block binding of MEM188 or CB56.